Få mere at vide
BOLA Tube Fittings
Artikelnummer. 10718062
Skift visning
Diameter (metrisk):
4 mm
6 mm
12 mm
14 mm
16 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Diameter (metrisk)
10718062 14 mm
10268590 4 mm
10349371 6 mm
10257591 12 mm
10525201 16 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 10718062 Leverandør BOLA Leverandørnr. D50316
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Tube Fittings PTFE, nut made of glass-fibre reinforced PTFE
  • Straight tube fitting made of PTFE with nuts made of glass-fibre reinforced PTFE.
  • Universal chemical resistance, the product is only exposed to PTFE.
Tekniske data
Certificeringer/compliance FDA
Diameter (metrisk) 14 mm
Autoklaverbar Autoclavable
Samlet længde (metrisk) 14 mm
Farve White
Fleksibilitet Rigid