missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubby Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99110AF700
Additional Details : Weight : 0.00970kg
Description
Tubby Polyclonal specifically detects Tubby in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Tubby | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
rd5, tubby (mouse) homolog, tubby homolog (mouse), tubby homologue, tubby protein homolog | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Tubby. | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuroscience, Transcription Factors and Regulators | |
7275 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |