missing translation for 'onlineSavingsMsg'
Learn More

Tubastatin A Hydrochloride, TRC

Product Code. 30010605
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30010605 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010605 Supplier Toronto Research Chemicals Supplier No. TRCT89701010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Tubastatin A Hydrochloride
CAS 1310693-92-5
Molekylær formel C20H22ClN3O2
Mængde 10 mg
Synonym N-Hydroxy-4-[(1,2,3,4-tetrahydro-2-methyl-5H-pyrido[4,3-b]indol-5-yl)methyl]benzamide Hydrochloride
InChI formel InChI=1S/C20H21N3O2.ClH/c1-22-11-10-19-17(13-22)16-4-2-3-5-18(16)23(19)12-14-6-8-15(9-7-14)20(24)21-25;/h2-9,25H,10-13H2,1H3,(H,21,24);1H
SMIL O=C(NO)C(C=C1)=CC=C1CN2C3=CC=CC=C3C4=C2CCN(C)C4.Cl
IUPAC navn N-hydroxy-4-[(2-methyl-3,4-dihydro-1H-pyrido[4,3-b]indol-5-yl)methyl]benzamide;hydrochloride
Molekylvægt (g/mol) 371.86
Formel vægt 371.1401
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.