Få mere at vide
Tocris Bioscience™ Tubastatin A hydrochloride
Potent HDAC6 inhibitor
Brand: Tocris Bioscience™ 6270/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Tubastatin A hydrochloride is a potent HDAC6 inhibitor (IC50 = 0.015 μM). Exhibits some selectively for HDAC6 over HDAC8 and 1 (IC50 values are 0.854 and 16.4 μM, respectively). Induces elevated levels of α-tubulin and protects against glutathione-induced oxidative stress in primary neuronal cell culture. Reverses the axonal loss in peripheral neurons in a mouse model of Charcot-Marie-Tooth disease.Tekniske data
| N-Hydroxy-4-[(1,2,3,4-tetrahydro-2-methyl-5H-pyrido[4,3-b]indol-5-yl)methyl]benzamide hydrochloride | |
| 1310693-92-5 | |
| >98% | |
| 10 mg |