missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies TTNPB

Artikelnummer. 17150142
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17150142 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17150142 Leverandør STEMCELL Technologies Leverandørnr. 72892

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Retinoid pathway activator; Activates retinoic acid receptor (RAR)

  • TTNPB is an analog of retinoic acid that potently and selectively activates retinoic acid receptors (RAR; EC50 = 21, 4, and 2.4 nM for RARα, RARβ, and RARγ, respectively; Beard et al.; Wong et al.)
  • In combination with CHIR99021 or Activin A, induces intermediate mesoderm formation from human or mouse pluripotent stem cells, respectively (Araoka et al.; Oeda et al.)
  • Promotes neuronal differentiation in cultured chick caudal neural plate explants (Diez del Corral et al.)
  • Enables chemical reprogramming (without genetic factors) of mouse embryonic fibroblasts to induced pluripotent stem (iPS) cells, in combination with CHIR99021, Tranylcypromine, VaLproic Acid, 3-Deazaneplanocin A, and E-616452 (Hou et al.)
  • Induces the in vitro growth and differentiation to granulocytes of myeloid progenitors isolated from myelodysplastic syndrome (MDS) patients (Fabian et al.)

Tekniske data

CAS 71441-28-6
Synonym AGN 191183; Arotinoid Acid; Ro 13-7410
Mængde 10 mg
Molekylær formel C24H28O2
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.