Learn More
TTC39C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564239
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TSFHTALELA VDQREIQHVC LYEIGWCSMI ELNFKDAFDS FERLKNESRW SQCYYAYLTA VCQGATGDVD GAQIVFKEVQ KLFKRKNNQI EQFSV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
TTC39C is a protein coding gene.Specifications
TTC39C | |
Polyclonal | |
Unconjugated | |
TTC39C | |
C18orf17; HsT2697; tetratricopeptide repeat domain 39C; tetratricopeptide repeat protein 39C; TPR repeat protein 39C; TTC39C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
125488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N584 | |
TTC39C | |
Recombinant protein corresponding to Human TTC39C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only