missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TSPAN1 Antibody (4/12), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-12061R
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
Tekniske data
| TSPAN1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 10103 | |
| Store at 4C in the dark. | |
| IgG κ |
| ELISA, Immunohistochemistry, Immunofluorescence | |
| 2022-04-12 00:00:00 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 | |
| This antibody was raised by immunising BALB/c mice with the major extracellular domain of tetraspanin 1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion