missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TSLP Antibody (55N1E3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80997
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TSLP Monoclonal specifically detects TSLP in Human samples. It is validated for Western Blot.
Tekniske data
| TSLP | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 0.5-2ug/ml | |
| thymic stromal lymphopoietin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 55N1E3 | |
| Unconjugated | |
| PBS | |
| TSLP | |
| amino acids (27-158) MDYDFTNCDFEkIkAAYLSTISkDLITYMSGTkSTEFNNTVSCSNRPHCLTEIQSLTFNP∽TAGCASLAkEMFAMkTkAALAIWCPGYSETQINATQAMkkRRkRkVTTNkCLEQVSQLQG∽LWRRFNRPLLkQQL | |
| 0.1 mg | |
| Biologically Active Proteins, Innate Immunity | |
| 85480 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion