missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSGA14 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-86015-0.1ML
Additional Details : Weight : 0.00970kg
Description
TSGA14 Polyclonal specifically detects TSGA14 in Human samples. It is validated for Western Blot.Specifications
TSGA14 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
95681 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
centrosomal protein 41 kDa, centrosomal protein of 41 kDa, Cep41, DKFZp762H1311, testis specific protein A14, testis specific, 14, Testis-specific gene A14 protein | |
The immunogen is a synthetic peptide directed towards the middle region of Human TSGA14 | |
0.1 mL | |
Primary | |
Human | |
Purified |