missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSG101 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-98994AF647
Additional Details : Weight : 0.00970kg
Description
TSG101 Polyclonal specifically detects TSG101 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
TSG101 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 647 | |
ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human TSG101. | |
0.1 mL | |
Autophagy, Cancer, Microautophagy | |
7251 | |
Store at 4°C in the dark. | |
IgG |