Få mere at vide
Thermo Scientific™ TSE Series Solid Door Laboratory Freezers
Artikelnummer. 31783322 Shop alle Thermo Scientific produkter
Skift visning
Kapacitet (metrisk):
77 L
266 L
564 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Kapacitet (metrisk)
31783322 564 L
31784000 77 L
31740920 77 L
31782587 266 L
31740936 266 L
31740923 564 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Artikelnummer. 31783322 Leverandør Thermo Scientific™ Leverandørnr. TSE2220FV
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Thermo Scientific TSE Series Laboratory Freezers deliver dependable cold storage for routine lab needs. Preset to -20°C, these auto-defrost units offer digital control and temperature alarms for easy, reliable use in practical undercounter and upright single door sizes.
Thermo Scientific TSE Series Laboratory Freezers provide dependable temperature performance and straightforward operation for routine lab use. Auto-defrost functionality, digital control, and essential features support everyday cold storage needs. All models use SNAP and F-Gas compliant hydrocarbon refrigerants and include a two-year warranty, offering practical value for daily laboratory workflows.
  • Designed for everyday research and academic lab applications with good performance and easy-to-use digital controls
  • Quiet operation, less than 53 dB
  • SNAP compliant green refrigerants
  • Auto defrost
  • Four 2 in. casters, front casters are lockable
  • Hi/Lo Temperature, and door ajar failure visual alarms
  • Rear access port
  • Keyed door locks, compatible with popular E-lock adapters
Stacking Kit accessory is available
TRUSTED_SUSTAINABILITY
Tekniske data
Type Upright
Kapacitet (engelsk) 20 cu.ft.
Kapacitet (metrisk) 564 L
Temperaturområde -15°C to -25°C
Temperatur (setpunkt) -20°C
Dør stil Solid
Antal døre 1
Skabsmateriale Painted Steel
Kammermateriale Painted Steel
Isolering HFC-free Polyurethane Foam
Udvendig finish White-coated Steel
Hylder 4
Certificeringer/compliance CE
Stil Upright
Vise Digital
Hjul Locking
Kontroltype Digital
Havne 1 Rear-access Port
garanti 2 Years Parts & Labor
Afrimning Auto
Belysning LED
Dimensioner (DxBxH) Udvendig 34.7 x 28.1 x 79.7 in. (88.1 x 71.3 x 202.6 cm)
Mål (D x B x H) Indvendig 27.6 x 23.5 x 59.3 in. (70.0 x 59.6 x 150.5 cm)
Overvågningsmuligheder Audible and Visual High and Low Temperature Alarms
Adgangssikkerhed Key Lock
Stiktype EU
Spænding 230 V
Frekvens 50 Hz
Energiforbrug 4.42 kw-hr/day
Kompressor Hermetic
Kølemiddel R290
Forsendelsesvægt (engelsk) 335 lb.
Forsendelsesvægt (metrisk) 152 kg