Få mere at vide
Tryptophol-d4, TRC
Artikelnummer. 30450268
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30450268 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450268 Leverandør Toronto Research Chemicals Leverandørnr. TRCT894702100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tryptophol-d4
CAS 75238-45-8
Renhedsgrad noter HPLC
Molekylær formel C10 D4 H7 N O
Mængde 100 mg
Synonym 1H-Indole-3-ethan-alpha,alpha,beta,beta-d4-ol
InChI formel InChI=1S/C10H11NO/c12-6-5-8-7-11-10-4-2-1-3-9(8)10/h1-4,7,11-12H,5-6H2/i5D2,6D2
SMIL [2H]C([2H])(O)C([2H])([2H])c1c[nH]c2ccccc12
IUPAC navn 1,1,2,2-tetradeuterio-2-(1H-indol-3-yl)ethanol
Molekylvægt (g/mol) 165.225
Formel vægt 165.109
Procent renhed >95