Få mere at vide
Tocris Bioscience™ TRPV3 74a
Beskrivelse
TRPV3 74a is a selective TRPV3 antagonist (KB = 0.56 μM). Displays no significant activity against a panel of other ion channels. Exhibits efficacy in animal models of neuropathic pain. Brain penetrant.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (αS)-α-[cis-3-Hydroxy-3-methyl-1-[4-(trifluoromethyl)-2-pyridinyl]cyclobutyl]-2-pyridinemethanol |
| CAS | 1432051-63-2 |
| Mængde | 5 mg |
| Mål | TRPV Antagonists |
| Molekylær formel | C17H17F3N2O2 |
| Formel vægt | 3434 |
| Renhed | 98% |