missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ TrpC2-like protein Recombinant Protein Antigen
Shop alle Bio Techne produkter
Klik for at se tilgængelige muligheder
Mængde:
100 μl
Beskrivelse
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human TrpC2-like protein. Source: E.coli Amino Acid Sequence: MAPVKISHVVSFSSQDPKYPVENLLNPDSPRRPWLGCPQDKSGQLKVELQLERAVPTGYIDVGNCGCAFLQ The TrpC2-like protein Recombinant Protein Antigen is derived from E. coli. The TrpC2-like protein Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Tekniske data
Tekniske data
| Gen-id (Entrez) | 100133315 |
| Oprensningsmetode | >80% by SDS-PAGE and Coomassie blue staining |
| Fælles navn | TrpC2-like protein Recombinant Protein Antigen |
| Indhold og opbevaring | Store at −20C. Avoid freeze-thaw cycles. |
| Formulering | PBS and 1M Urea, pH 7.4. |
| Til brug med (applikation) | Blocking/Neutralizing, Control |
| Gene Alias | LOC100133315 transient receptor potential cation channel, subfamily C, member 2-like |
| Gen symbol | LOC100133315 |
| Etikettype | Unlabeled |
| Produkttype | Recombinant Protein Antigen |
| Vis mere |
For Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion