Få mere at vide
Troxipide, TRC
Artikelnummer. 30418350
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418350 100 mg
30415731 250 mg
30423878 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30418350 Leverandør Toronto Research Chemicals Leverandørnr. TRCT011240100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 30751-05-4
Molekylær formel C15H22N2O4
Molekylvægt (g/mol) 294.35
Synonym KU 54, 3-(3,4,5-Trimethoxybenzamido)piperidine, 3,4,5-Trimethoxy-N-3-piperidinylbenzamide, Aplace
IUPAC navn 3,4,5-trimethoxy-N-piperidin-3-ylbenzamide
SMIL COc1cc(cc(OC)c1OC)C(=O)NC2CCCNC2
Tekniske data
Kemisk navn eller materiale Troxipide
Mængde 100 mg
InChI formel InChI=1S/C15H22N2O4/c1-19-12-7-10(8-13(20-2)14(12)21-3)15(18)17-11-5-4-6-16-9-11/h7-8,11,16H,4-6,9H2,1-3H3,(H,17,18)
Formel vægt 294.158