Learn More
Troponin T (Cardiac Isoform) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Troponin T, Cardiac Isoform Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
1065.00 DKK - 6475.00 DKK
Specifications
Antigen | Troponin T, Cardiac Isoform Ab-1 |
---|---|
Clone | 13-11 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12660847
|
Epredia
MS-295-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12680847
|
Epredia
MS-295-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12699030
|
Epredia
MS-295-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12670847
|
Epredia
MS-295-P1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12650847
|
Epredia
MS-295-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12620857
|
Epredia
MS-295-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Troponin T is a thin filament regulatory protein of striated muscle that binds to troponin C and I and to tropomyosin and is required for calcium-dependent ATPase activity of myofibrillar proteins. It is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. This expression in the human heart is affected by heart failure.Host Species: Mouse
Clone: 13-11
Isotype: IgG1
Species Reactivity: Human, Pig, Dog, Rabbit, Rat, Mouse, Guinea Pig, Chicken, Sparrow and Goldfish. Does not react with frog.
Epitope: FMPNLVPPKI (5' region of the cardiac gene exon 10)
Immunogen: Purified rabbit cardiac troponin T isoform (TnT4R)
Positive Control: Cardiac muscle
Cellular Localization: Mostly over the I bands in myofibrils Ab-1 specifically recognizes the cardiac isoform of Troponin T (cTnT).
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Electron Microscopy
Specifications
Troponin T, Cardiac Isoform Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Canine, Chicken, Goldfish, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Sparrow | |
IgG1 |
13-11 | |
Monoclonal | |
Mouse | |
Muscle Biology | |
Purified rabbit cardiac troponin T isoform (TnT4R) | |
Primary |