Learn More
Alfa Aesar™ Tropine DL-tropate, 99%
Reduces brain levels of acetylcholine
Brand: Alfa Aesar™ J64677.03
Additional Details : CAS Number : 51-55-8 Weight : 0.00100kg Transport : UN number : 1544 Chem class : 6.1 Pack group : II
Specifications
1g | |
Powder | |
0.99 | |
C17H23NO3 | |
DL-Hyoscyamine; Atropine | |
Signal Transduction Reagent | |
Powder |
White | |
Reduces brain levels of acetylcholine | |
51-55-8 | |
289.37g/mol | |
Light sensitive | |
91260 |