Få mere at vide
Trofosfamide-d4, TRC
Artikelnummer. 30453084
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30453084 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453084 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8920022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trofosfamide-d4
CAS 1189884-36-3
Molekylær formel C9H14D4Cl3N2O2P
Mængde 2.5 mg
InChI formel InChI=1S/C9H18Cl3N2O2P/c10-2-6-13-5-1-9-16-17(13,15)14(7-3-11)8-4-12/h1-9H2/i1D2,5D2
SMIL [2H]C1([2H])COP(=O)(N(CCCl)CCCl)N(CCCl)C1([2H])[2H]
IUPAC navn N,N,3-tris(2-chloroethyl)-4,4,5,5-tetradeuterio-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine
Molekylvægt (g/mol) 327.61
Formel vægt 326.0423