Få mere at vide
Tocris Bioscience™ Trofinetide
Beskrivelse
Trofinetide is an IGF-1 synthetic analog. It suppresses expression of IL-1β, TNF-α, IFN-γ and E-selectin, and inhibits both acute and delayed neuroinflammation following penetrating ballistic-like brain injury (PBBI) in rats.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Glycyl-2-methyl-L-prolyl-L-glutamic acid |
| CAS | 853400-76-7 |
| Mængde | 10 mg |
| Mål | IGF-I/IGF-1 |
| Molekylær formel | C13H21N3O6 |
| Renhed | 0.98 |