Få mere at vide
Trofinetide (>90%), TRC
Artikelnummer. 30455963
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30455963 2.5 mg 2.5mg
30449314 0.5 mg -
30452833 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30455963 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8925252.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 853400-76-7
Molekylær formel C13H21N3O6
Molekylvægt (g/mol) 315.32
Synonym Glycyl-2-methyl-L-prolyl-L-glutamic Acid, Glycyl-L-2-methylprolyl-L-glutamic Acid, NNZ 2566
IUPAC navn (2S)-2-[[(2S)-1-(2-aminoacetyl)-2-methylpyrrolidine-2-carbonyl]amino]pentanedioic acid
SMIL C[C@]1(CCCN1C(=O)CN)C(=O)N[C@@H](CCC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Trofinetide
Mængde 2.5 mg
InChI formel InChI=1S/C13H21N3O6/c1-13(5-2-6-16(13)9(17)7-14)12(22)15-8(11(20)21)3-4-10(18)19/h8H,2-7,14H2,1H3,(H,15,22)(H,18,19)(H,20,21)/t8-,13-/m0/s1
Formel vægt 315.143