Learn More
TRMT61B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555402
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KRGTAITFPK DINMILSMMD INPGDTVLEA GSGSGGMSLF LSKAVGSQGR VISFEVRKDH HDLAKKNYKH WRDSWKLSHV EEWPDNVDFI HKDISGATED IKSLTFDAVA LDMLN Highest antigen sequence identity to the following orthologs: Mouse - 22%, Rat - 24%.
TRMT61B Polyclonal antibody specifically detects TRMT61B in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
TRMT61B | |
Polyclonal | |
Unconjugated | |
TRMT61B | |
1110039F03Rik; 623451; Gm6431; mRNA methyladenosine-N(1)-methyltransferase; potential tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61B; potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B; TRMT61B; tRNA (adenine(58)-N(1))-methyltransferase, mitochondrial; tRNA methyltransferase 61 homolog B; tRNA methyltransferase 61B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55006 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BVS5 | |
TRMT61B | |
Recombinant protein corresponding to Human TRMT61B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only