missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkA Antibody (MNAC13), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81074C
Additional Details : Weight : 0.00970kg
Description
TrkA Monoclonal specifically detects TrkA in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
TrkA | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
NTRK1 | |
Human TrkA ECD. | |
0.1 mL | |
Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase | |
4914 | |
Human | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
MNAC13 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody binds to the extracellular domain of TrkA. | |
Store at 4°C in the dark. | |
IgG κ |