missing translation for 'onlineSavingsMsg'
Learn More
Tritolyl phosphate, mixture of isomers Product Code.: 11448017

Tritolyl phosphate, mixture of isomers

Product Code. p-7023648
500 mL, 500mL
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 11448017

Brand: Thermo Scientific Alfa Aesar A17433.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 1330-78-5 | C21H21O4P

Tritolyl phosphate, mixture of isomers is used as plasticizer in nitrocellulose, acrylate lacquers, and in polyvinyl chloride. It is a fire retardant for plastics and rubbers. air filter medium, solvent mixtures and heat exchange medium. It is employed as a gasoline additive and as a lead scavenger for tetraethyllead. Further, it acts as a solvent for extraction, especially for nitrocellulose and polymers. In addition to this, it is used for the waterproofing of materials, an antiwear and extreme pressure additive in lubricants and hydraulic fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tritolyl phosphate, mixture of isomers is used as plasticizer in nitrocellulose, acrylate lacquers, and in polyvinyl chloride. It is a fire retardant for plastics and rubbers. air filter medium, solvent mixtures and heat exchange medium. It is employed as a gasoline additive and as a lead scavenger for tetraethyllead. Further, it acts as a solvent for extraction, especially for nitrocellulose and polymers. In addition to this, it is used for the waterproofing of materials, an antiwear and extreme pressure additive in lubricants and hydraulic fluids.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, sulfuric acid, caustics, ammonia, aliphatic amines, alkanolamines, isocyanates, alkylene oxides, epichlorohydrin and nitromethane.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-78-5
Molecular Formula C21H21O4P
MDL Number MFCD02102116
Synonym Tricresyl phosphate; TCP

Specifications

Density 1.167
Boiling Point 265°C (10 mmHg)
Flash Point 250°C (482°F)
Odor Odorless
Linear Formula (CH3C6H4O)3P=O
Quantity 500 mL
UN Number UN2574
Merck Index 14,9763
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 368.37
Percent Purity 98%
Chemical Name or Material Tritolyl phosphate, mixture of isomers, ortho-isomers <1%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10254391 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeA17433.AP,A17433.0F
Lot Number10250481 Certificate TypeCertificate of Analysis Date16/04/2024 Product CodeA17433.AP,A17433.0F
Lot Number10239355 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeA17433.AP,A17433.0F
Lot Number10121676 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA17433.AP,A17433.0F
Lot Number10178302 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA17433.AP,A17433.0F

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tritolyl phosphate, mixture of isomers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.