missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
BD Biosciences™ Tritest: CD3 FITC/CD4 PE/CD45 PerCP (CE/IVD)
Klik for at se tilgængelige muligheder
Mængde:
50 tests
Beskrivelse
- Clone SK7 (Leu-4) SK3 (Leu3a) 2D1 (CE/IVD)
- This BD Tritest™ CD3 fluorescein isothiocyanate (FITC)/CD4 phycoerythrin (PE)/CD45 peridinin chlorophyll protein (PerCP) is a three-color direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of mature human T-lymphocytes (CD3+) and helper/inducer (CD3+CD4+) T-lymphocytes subsets in erythrocyte-lysed whole blood.
- When used with BD Trucount™ Tubes, absolute counts of these populations can be enumerated from a single tube.
- This BD Tritest reagent and Trucount tubes can be used with the FACS™ Loader. The reagent can be used with or without an isotype control.
- Contents:
- Specificity: CD3, Clone: SK7 (Leu-4), Format: FITC, Isotype: IgG<sub>1</sub> K, Entrez Gene ID: 916
- Specificity: CD4, Clone SK3 (Leu3a), Format: PE, Isotype: IgG<sub>2b</sub> K, Entrez Gene ID: 920
- Specificity: CD45, Clone: 2D1, Format: PerCP, Isotype: IgG<sub>1</sub> K
Tekniske data
Tekniske data
| Indhold og opbevaring | Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration. |
| Antal prøver | 50 Tests |
| Mængde | 50 tests |
| Regulatorisk status | CE/IVD |
| Produkttype | Immunofluorescence Flow Cytometry Test Reagents |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion