Få mere at vide
Tris(Pentafluorophenyl)borane Hydrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tris(Pentafluorophenyl)borane Hydrate |
| CAS | 1246935-88-5 |
| Molekylær formel | C18BF15 •H2O |
| Mængde | 2.5 g |
| Synonym | Tris(2,3,4,5,6-pentafluorophenyl)borane, BF 15, Perfluorotriphenylboron Hydrate, Perfluorotrisphenylborane, Tri(pentafluorophenyl)boron Hydrate, Tri(perfluorophenyl)borane Hydrate, Tris(pentafluorophenyl)borane, Tris(pentafluorophenyl)boron Hydrate, Tris(perfluorophenyl)borane Hydrate, Tris(perfluorophenyl)boron Hydrate |
| InChI formel | InChI=1S/C18BF15.H2O/c20-4-1(5(21)11(27)16(32)10(4)26)19(2-6(22)12(28)17(33)13(29)7(2)23)3-8(24)14(30)18(34)15(31)9(3)25;/h;1H2 |
| SMIL | FC1=C(F)C(F)=C(F)C(F)=C1B(C2=C(F)C(F)=C(F)C(F)=C2F)C3=C(F)C(F)=C(F)C(F)=C3F.O |
| IUPAC navn | tris(2,3,4,5,6-pentafluorophenyl)borane;hydrate |
| Molekylvægt (g/mol) | 511.98 |
| Formel vægt | 529.9959 |