missing translation for 'onlineSavingsMsg'
Learn More

Tris(Hydroxymethyl) Methylamine, Biological Buffer, Fisher Chemical™

Catalog Number p-7073646
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
4679
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
4679
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
Melting Point 168.5°C
Boiling Point 219°C
Color White
pH 10.4
Physical Form Powder or Solid
Quantity 100 g
Packaging Plastic powder jar
Formula Weight 121.14g/mol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'