missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 20X), with 1% Tween 20

Catalog Number p-7042763
Click to view available options
Quantity:
1 L
2 L
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
MFCD00678198
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
MFCD00678198
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration 20X
Physical Form Liquid
Quantity 1 L
Chemical Name or Material TRIS-buffered saline, with 0.5% Tween 20

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'