missing translation for 'onlineSavingsMsg'
Learn More

Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog Number p-3523619
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
80049-61-2
C11H14F6N3PRu
434.29
MFCD02684569
80049-61-2
C11H14F6N3PRu
434.29
MFCD02684569

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown or Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Ru)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: slightly soluble.
Formula Weight 434.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate
compliance-icons
Product Identifier
  • Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'