Få mere at vide
Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl) Isocyanurate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Irganox 1790 |
| CAS | 40601-76-1 |
| Molekylær formel | C42 H57 N3 O6 |
| Mængde | 10 g |
| Synonym | 1,3,5-Tris[[4-(1,1-dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]methyl]-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Tri(4-tert-butyl-2,6-dimethyl-3-hydroxybenzyl)-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Tris(2,6-dimethyl-3-hydroxy-4-tert-butylbenzyl) isocyanurate, 1,3,5-Tris(2,6-dimethyl-4-tert-butyl-3-hydroxybenzyl) isocyanurate, 1,3,5-Tris(4-tert-butyl-2,6-dimethyl-3-hydroxybenzyl) isocyanurate, 1,3,5-Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl) isocyanurate, 1,3,5-Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl)-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl)-1,3,5-triazine-2,4,6-trione, 1,3,5-Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl)-s-triazine-2,4,6-(1H,3H,5H)-trione, 1,3,5-Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl)isocyanuric acid, 1,3,5-Tris(4-tert-butyl-3-hydroxy-2,6-dimethylphenylmethyl)-1,3,5-triazine-2,4,6-(1H,3H,5H)trione, 1,3,5-Tris[(4-tert-butyl-3-hydroxy-2,6-xylyl)methyl]-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, ADK-ARKLS DH 48, AO 1790, Antioxidant 1790, Antioxidant 1970, Antioxidant CY, CN 1790, CY 1790, Chinox 1790, Cyanox 1790, Cyanox 1970, Cyanox CY 1790, Cynomix 1790, DN 90, Irganox 1790, Irganox 3790, Lowinox 1790, Songnox 1790, TS 274, Thanox 1790, Tris(3-hydroxy-4-tert-butyl-2,6-dimethylbenzyl) isocyanurate, Tris(4-tert-butyl-2,6-dimethyl-3-hydroxybenzyl) isocyanurate, Tris(4-tert-butyl-3-hydroxy-2,6-dimethylbenzyl) isocyanurate, Tris[(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)methyl]-1,3,5-triazine-2,4,6-trione |
| InChI formel | InChI=1S/C42H57N3O6/c1-22-16-31(40(7,8)9)34(46)25(4)28(22)19-43-37(49)44(20-29-23(2)17-32(41(10,11)12)35(47)26(29)5)39(51)45(38(43)50)21-30-24(3)18-33(42(13,14)15)36(48)27(30)6/h16-18,46-48H,19-21H2,1-15H3 |
| SMIL | Cc1cc(c(O)c(C)c1CN2C(=O)N(Cc3c(C)cc(c(O)c3C)C(C)(C)C)C(=O)N(Cc4c(C)cc(c(O)c4C)C(C)(C)C)C2=O)C(C)(C)C |
| IUPAC navn | 1,3,5-tris[(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)methyl]-1,3,5-triazinane-2,4,6-trione |
| Molekylvægt (g/mol) | 699.92 |
| Formel vægt | 699.4247 |