Learn More
Tris(4-methoxyphenyl)phosphine, 95%, ACROS Organics™
Brand: Acros Organics 422240100
Additional Details : CAS Number : 855-38-9 Weight : 0.06000kg
Packaging | Glass bottle |
---|---|
Quantity | 10g |
Chemical Identifiers
855-38-9 | |
352.36 | |
UYUUAUOYLFIRJG-UHFFFAOYSA-N | |
70071 | |
COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC |
C21H21O3P | |
MFCD00014896 | |
tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine | |
tris(4-methoxyphenyl)phosphane |
Specifications
Tris(4-methoxyphenyl)phosphine | |
Glass bottle | |
131.0°C to 134.0°C | |
95% | |
94% min. (GC) | |
MFCD00014896 | |
Solubility in water: practically insoluble. Other solubilities: soluble in methanol | |
COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC | |
352.36 | |
352.36 | |
95% |
Authentic | |
White to Yellow | |
10g | |
855-38-9 | |
C21H21O3P | |
tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine | |
UYUUAUOYLFIRJG-UHFFFAOYSA-N | |
tris(4-methoxyphenyl)phosphane | |
70071 | |
Crystalline Powder |