Få mere at vide
Tris(3-sulfophenyl)phosphine Trisodium (~85%), TRC
Artikelnummer. 30456658
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456658 10 g
30455179 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456658 Leverandør Toronto Research Chemicals Leverandørnr. TRCT89593810G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tris(3-sulfophenyl)phosphine Trisodium
CAS 63995-70-0
Molekylær formel C18H12Na3O9PS3
Mængde 10 g
Synonym Triphenylphosphine-m-trisulfonic Acid trisodium Salt
InChI formel InChI=1S/C18H15O9PS3.3Na/c19-29(20,21)16-7-1-4-13(10-16)28(14-5-2-8-17(11-14)30(22,23)24)15-6-3-9-18(12-15)31(25,26)27;;;/h1-12H,(H,19,20,21)(H,22,23,24)(H,25,26,27);;;/q;3*+1/p-3
SMIL C1=CC(=CC(=C1)S(=O)(=O)[O-])P(C2=CC(=CC=C2)S(=O)(=O)[O-])C3=CC(=CC=C3)S(=O)(=O)[O-].[Na+].[Na+].[Na+]
IUPAC navn trisodium;3-bis(3-sulfonatophenyl)phosphanylbenzenesulfonate
Molekylvægt (g/mol) 568.42
Formel vægt 567.9074