Få mere at vide
Tris(2-isopropylphenyl) Phosphate, TRC
Artikelnummer. 30450945
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30450945 2.5 g 2.5g
30450292 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30450945 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8853002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64532-95-2
Molekylær formel C27 H33 O4 P
Molekylvægt (g/mol) 452.52
Synonym Tris[2-(2-methylethyl)phenyl] Phosphate, Phenol, 2-(1-methylethyl)-, phosphate (3:1) (9CI), o-Cumenyl phosphate (6CI), Tris(2-isopropylphenyl) phosphate, Tris(o-isopropylphenyl) phosphate, Phenol, 2-(1-methylethyl)-, 1,1',1' '-phosphate
IUPAC navn tris(2-propan-2-ylphenyl) phosphate
SMIL CC(C)c1ccccc1OP(=O)(Oc2ccccc2C(C)C)Oc3ccccc3C(C)C
Tekniske data
Kemisk navn eller materiale tris(2-isopropylphenyl) Phosphate
Mængde 2.5 g
InChI formel InChI=1S/C27H33O4P/c1-19(2)22-13-7-10-16-25(22)29-32(28,30-26-17-11-8-14-23(26)20(3)4)31-27-18-12-9-15-24(27)21(5)6/h7-21H,1-6H3
Formel vægt 452.2116