Få mere at vide
Tris(2-chloroethyl)phosphate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tris-(2-chloroethyl)phosphate |
| CAS | 115-96-8 |
| Molekylær formel | C6 H12 Cl3 O4 P |
| Mængde | 1 g |
| Synonym | Tris-(2-chloroethyl)phosphate, Ethanol','2-chloro-','1,1',1''-phosphate, Ethanol','2-chloro-','phosphate (3:1) (8CI,9CI), 3CF, Amgard TCEP, CEF, CLP, Celluflex CEF, Disflamoll TCA, Fyrol CEF, Fyrol CF, Genomoll P, NSC 3213, Niax 3CF, Niax Flame Retardant 3CF, Phosphoric acid tris(2-chloroethyl) ester, Roflam E, TCEP, Tri(2-chloroethyl) phosphate, Tri(chloroethyl) phosphate, Tri(β-chloroethyl) phosphate, Tris(2-chloroethyl) orthophosphate, Tris(2-chloroethyl) phosphate, Tris(chloroethyl) phosphate, Tris(β-chloroethyl) phosphate |
| InChI formel | InChI=1S/C6H12Cl3O4P/c7-1-4-11-14(10,12-5-2-8)13-6-3-9/h1-6H2 |
| SMIL | ClCCOP(=O)(OCCCl)OCCCl |
| IUPAC navn | tris(2-chloroethyl) phosphate |
| Molekylvægt (g/mol) | 285.49 |
| Formel vægt | 283.9539 |