missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals

Catalog Number p-7034197
Click to view available options
Quantity:
5 mL
25 mL
100 mL
51805-45-9
C9H12O6P
247.16
MFCD00145469
PZBFGYYEXUXCOF-UHFFFAOYSA-K
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
2734570
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
51805-45-9
C9H12O6P
247.16
MFCD00145469
PZBFGYYEXUXCOF-UHFFFAOYSA-K
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
2734570
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O

Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.

Solubility
Miscible with water and methanol.

Notes
Air sensitive. Store in cool place. Incompatible with strong oxidizing agents
TRUSTED_SUSTAINABILITY
Linear Formula (HO2CCH2CH2)3P·HCl
Quantity 5 mL
UN Number UN3265
Solubility Information Miscible with water and methanol.
Formula Weight 286.65
Percent Purity 95%
Concentration or Composition (by Analyte or Components) 0.5M soln. in water
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'