Få mere at vide
Tris(1-chloro-2-propyl) Phosphate-d18, TRC
Artikelnummer. 30456887
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30456887 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456887 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8878621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tris(1-chloro-2-propyl) Phosphate-d18
CAS 1447569-78-9
Molekylær formel C9D18Cl3O4P
Mængde 1 mg
Synonym 1-Chloro-2-propanol-d6 2,2',2''-phosphate, TCPP-d18, Tris(1-chloro-2-propyl-1,2,2,3,3,3-d6) phosphate, Tris[2-chloro-1-(methyl-d3)ethyl-1,2,2-d3] phosphate, Tris(2-chloroisopropyl) phosphate-d18, Tris(2-chloroisopropyl) phosphate D18, Tris(chloroisopropyl) phosphate-d18, Tris(β-chloroisopropyl) phosphate-d18, Tris(1-chloro-2-propyl) phosphate-d18
InChI formel InChI=1S/C9H18Cl3O4P/c1-7(4-10)14-17(13,15-8(2)5-11)16-9(3)6-12/h7-9H,4-6H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D,8D,9D
SMIL [2H]C([2H])([2H])C([2H])(OP(=O)(OC([2H])(C([2H])([2H])[2H])C([2H])([2H])Cl)OC([2H])(C([2H])([2H])[2H])C([2H])([2H])Cl)C([2H])([2H])Cl
Molekylvægt (g/mol) 345.68
Formel vægt 344.1138