missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS, 0.5M buffer soln., pH 9.5, Thermo Scientific™
Click to view available options
Quantity:
500 mL
1 L
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Concentration | 0.5 M |
pH | 9.5 |
Physical Form | Liquid |
Quantity | 500 mL |
Chemical Name or Material | TRIS buffer soln. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'