missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TRIS, 0.5M buffer soln., pH 9.0, Thermo Scientific™
Klik for at se tilgængelige muligheder
Mængde:
500 mL
1 L
Pakningsstørrelse:
1 L
500ml
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away fom strong oxidizing agents.
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.
Solubility
Soluble in water and organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away fom strong oxidizing agents.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | TRIS buffer soln. |
| Koncentration | 0.5 M |
| pH | 9.0 |
| Fysisk form | Liquid |
| Mængde | 500 mL |
| Opløselighedsinformation | Soluble in water and organic solvents. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion