missing translation for 'onlineSavingsMsg'
Learn More

TRIS, 0.5M buffer soln., pH 7.0, Thermo Scientific™

Catalog Number p-7042053
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Concentration 0.5 M
pH 7.0
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material TRIS buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'