Få mere at vide
Triprolidine Hydrochloride Monohydrate, Mikromol™
Beskrivelse
Triprolidine Hydrochloride Monohydrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Triprolidine Hydrochloride Monohydrate |
| CAS | 6138-79-0 |
| Analyt- eller komponentnavne | Triprolidine Hydrochloride Monohydrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H22 N2 . H Cl . H2 O |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Pyridine, 2-[(1E)-1-(4-methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]-, monohydrochloride, monohydrate, Pyridine, 2-[1-(4-methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]-, monohydrochloride, monohydrate, (E)-, Pyridine, 2-[3-(1-pyrrolidinyl)-1-p-tolylpropenyl]-, monohydrochloride, monohydrate, stereoisomer, Triprolidine hydrochloride monohydrate |
| InChI formel | InChI=1S/C19H22N2.ClH.H2O/c1-16-7-9-17(10-8-16)18(19-6-2-3-12-20-19)11-15-21-13-4-5-14-21;;/h2-3,6-12H,4-5,13-15H2,1H3;1H;1H2/b18-11+;; |
| SMIL | O.Cl.Cc1ccc(cc1)\C(=C/CN2CCCC2)\c3ccccn3 |
| IUPAC navn | 2-[(E)-1-(p-tolyl)-3-pyrrolidin-1-yl-prop-1-enyl]pyridine;hydrate;hydrochloride |
| Vis mere |