missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripotassium citrate monohydrate, 97%, Thermo Scientific Chemicals
179.35 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2500 g
Description
It is used in the laboratory as a buffering agent and chelator. In renal function studies, potassium citrate is used as an alkanizing agent where the citric acid is a metabolic substrate in renal cells. Studies have shown that potassium citrate may increase bone density, it is therefore used in research experiments related to the treatment of osteoporosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 230°C (decomposition) |
Density | 1.98 g/mL |
Quantity | 500 g |
Merck Index | 14,7623 |
Solubility Information | Soluble in water (640mg/ml at 25°C),and glycerol. Insoluble in alcohol. |
Formula Weight | 324.42 (306.41 Anhydrous) |
Percent Purity | 97% |
Physical Form | Lump or Granular |
Chemical Name or Material | Tripotassium citrate monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'