missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar
Carry out environmental monitoring in your most regulated environments using Thermo Scientific™ Triple Wrapped Irradiated plates, for passive and active air sampling applications.
2295.00 DKK
Specifications
Color | Ivory, transparent |
---|---|
Content And Storage | 2 to 25°C |
Description | Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar |
Manufacturing Origin | Germany |
Product Type | Agar |
Product Code | Brand | Description | Quantity | Manufacturing Origin | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Manufacturing Origin | Price | Quantity & Availability | |||
15486030
|
Thermo Scientific™
PO5512D |
Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar | 100/Pk. | Germany |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Sabouraud Dextrose Agar settling plates, complete with VHP Indicator.
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.Three protective layers reduce risk of contamination:
- Primary film with moisture control patch
- Secondary film VHP barrier with immobilized desiccant sachet
- Tertiary film dust cover
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
Specifications
Ivory, transparent | |
Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar | |
Agar | |
100/Pk. |
2 to 25°C | |
Germany | |
Triple Wrapped - Irradiated |