Få mere at vide
Triphthazine Sulfoxide, TRC
Artikelnummer. 30451699
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30451699 25 mg
1 options
Artikelnummer. 30451699 Leverandør Toronto Research Chemicals Leverandørnr. TRCT72482025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trifluoperazine Sulfoxide
CAS 1549-88-8
Molekylær formel C21 H24 F3 N3 O S
Mængde 25 mg
Synonym 10-[3-(4-Methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-10H-phenothiazine 5-Oxide, 10-[3-(4-Methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)phenothiazine 5-Oxide, Trifluoperazine S-oxide, Trifluoperazine sulfoxide, Trifluoperazine sulphoxide, Trifluoroperazine sulfoxide, Triphthazine sulfoxide
InChI formel InChI=1S/C21H24F3N3OS/c1-25-11-13-26(14-12-25)9-4-10-27-17-5-2-3-6-19(17)29(28)20-8-7-16(15-18(20)27)21(22,23)24/h2-3,5-8,15H,4,9-14H2,1H3
SMIL CN1CCN(CCCN2c3ccccc3S(=O)c4ccc(cc24)C(F)(F)F)CC1
IUPAC navn 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine 5-oxide
Molekylvægt (g/mol) 423.5
Formel vægt 423.1592