Få mere at vide
Triphenylene-d12, CDN
Artikelnummer. 30319856
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319856 0.1 g
30310265 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319856 Leverandør CDN Leverandørnr. CDND00410.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triphenylene-D12
CAS 17777-56-9
Molekylær formel C18 2H12
Mængde 0.1 g
Synonym Triphenylene-d12 (7 CI,8 CI,9 CI), Triphenylene D12
InChI formel InChI=1 S/C18H12/c1-2-8-14-13(7-1)15-9-3-4-11-17(15)18-12-6-5-10-16(14)18/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D
SMIL [2 H]c1c([2 H])c([2 H])c2c3c([2 H])c([2 H])c([2 H])c([2 H])c3c4c([2 H])c([2 H])c([2 H])c([2 H])c4c2c1[2 H]
IUPAC navn 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriotriphenylene
Molekylvægt (g/mol) 240.36
Formel vægt 240.1692 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity