Få mere at vide
Triphenyl-d15 Phosphate, CDN
Artikelnummer. 30315947
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315947 0.25 g -
30322081 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315947 Leverandør CDN Leverandørnr. CDND74700.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triphenylphosphate D15
CAS 1173020-30-8
Molekylær formel C18 2H15 O4 P
Mængde 0.25 g
Synonym Phen-2,3,4,5,6-d5-ol, 1,1',1''-phosphate, Triphenylphosphate D15
InChI formel InChI=1 S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D,15 D
SMIL [2 H]c1c([2 H])c([2 H])c(OP(=O)(Oc2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])Oc3c([2 H])c([2 H])c([2 H])c([2 H])c3[2 H])c([2 H])c1[2 H]
IUPAC navn tris(2,3,4,5,6-pentadeuteriophenyl) phosphate
Molekylvægt (g/mol) 341.38
Formel vægt 341.1649 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity