missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripelennamine N-Oxide, Mikromol™
Description
Tripelennamine N-Oxide
Specifications
Specifications
| Kemisk navn eller materiale | Tripelennamine N-Oxide |
| CAS | 60317-19-3 |
| Analyt- eller komponentnavne | Tripelennamine N-Oxide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H21 N3 O |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1,2-Ethanediamine, N1,N1-dimethyl-N2-(phenylmethyl)-N2-2-pyridinyl-, N1-oxide, 1,2-Ethanediamine, N,N-dimethyl-N'-(phenylmethyl)-N'-2-pyridinyl-, N-oxide (9CI), Tripelennamine, N-oxide (6CI) |
| InChI formel | InChI=1S/C16H21N3O/c1-19(2,20)13-12-18(16-10-6-7-11-17-16)14-15-8-4-3-5-9-15/h3-11H,12-14H2,1-2H3 |
| SMIL | C[N+](C)([O-])CCN(Cc1ccccc1)c2ccccn2 |
| IUPAC navn | 2-[benzyl(pyridin-2-yl)amino]-N,N-dimethylethanamine oxide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?