Få mere at vide
Tripelennamine Hydrochloride, Mikromol™
Artikelnummer. 17942126
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
17942126 250 mg
1 options
Artikelnummer. 17942126 Leverandør Mikromol Leverandørnr. MM1631.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tripelennamine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tripelennamine hydrochloride
CAS 154-69-8
Analyt- eller komponentnavne Tripelennamine hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C16 H21 N3 . Cl H
Certificeringer/compliance ISO 17034
Synonym 1,2-Ethanediamine, N,N-dimethyl-N'-(phenylmethyl)-N'-2-pyridinyl-, monohydrochloride, 1,2-Ethanediamine, N1,N1-dimethyl-N2-(phenylmethyl)-N2-2-pyridinyl-, hydrochloride, Pyridine, 2-[benzyl[2-(dimethylamino)ethyl]amino]-, monohydrochloride, 2-[Benzyl[2-(dimethylamino)ethyl]amino]pyridine hydrochloride, Azaron, Dehistin, Dehistin monohydrochloride, N,N-Dimethyl-N'-(2-pyridyl)-N'-benzylethylenediamine hydrochloride, N-Benzyl-N',N'-dimethyl-N-2-pyridylethylenediamine hydrochloride, NIH 10186, PBZ, Piristin, Pyribenzamine hydrochloride, Pyribenzamine monohydrochloride, Pyrinamine, ReCovr, Stanzamine, Tripelennamine hydrochloride, Tripelennamine monohydrochloride, Vetibenzamine, Vetobenzamina
InChI formel InChI=1S/C16H21N3.ClH/c1-18(2)12-13-19(16-10-6-7-11-17-16)14-15-8-4-3-5-9-15;/h3-11H,12-14H2,1-2H3;1H
SMIL Cl.CN(C)CCN(Cc1ccccc1)c2ccccn2
IUPAC navn N'-benzyl-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;hydrochloride
Molekylvægt (g/mol) 291.82
Mængde 250 mg
Formel vægt 291.1502 g/mol