Få mere at vide
Tripelennamine Hydrochloride, LoGiCal
Artikelnummer. 30463965
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463965 10 mg
1 options
Artikelnummer. 30463965 Leverandør LoGiCal Leverandørnr. LGCFOR1631.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tripelennamine hydrochloride
CAS 154-69-8
Analyt- eller komponentnavne Tripelennamine hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C16 H21 N3 . Cl H
Certificeringer/compliance ISO 9001
Synonym 1,2-Ethanediamine, N,N-dimethyl-N'-(phenylmethyl)-N'-2-pyridinyl-, monohydrochloride;1,2-Ethanediamine, N1,N1-dimethyl-N2-(phenylmethyl)-N2-2-pyridinyl-, hydrochloride, Pyridine, 2-[benzyl[2-(dimethylamino)ethyl]amino]-, monohydrochloride, 2-[Benzyl[2-(dimethylamino)ethyl]amino]pyridine hydrochloride, Azaron, Dehistin, Dehistin monohydrochloride, N,N-Dimethyl-N'-(2-pyridyl)-N'-benzylethylenediamine hydrochloride, N-Benzyl-N',N'-dimethyl-N-2-pyridylethylenediamine hydrochloride, NIH 10186, PBZ, Piristin, Pyribenzamine hydrochloride, Pyribenzamine monohydrochloride, Pyrinamine, ReCovr, Stanzamine, Tripelennamine hydrochloride, Tripelennamine monohydrochloride, Vetibenzamine, Vetobenzamina
InChI formel InChI=1S/C16H21N3.ClH/c1-18(2)12-13-19(16-10-6-7-11-17-16)14-15-8-4-3-5-9-15;/h3-11H,12-14H2,1-2H3;1H
Løsningstype Neat
SMIL Cl.CN(C)CCN(Cc1ccccc1)c2ccccn2
IUPAC navn N'-benzyl-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 291.82
Mængde 10 mg
Formel vægt 291.1502 g/mol