Få mere at vide
Tripelennamine hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30325628
Skift visning
Mængde:
1 ml
Artikelnummer. Mængde
30325628 1 ml
1 options
Artikelnummer. 30325628 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA17892500AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tripelennamine hydrochloride
CAS 154-69-8
Analyt- eller komponentnavne Tripelennamine hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C16 H21 N3 . Cl H
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17034
Synonym 1,2-Ethanediamine, N,N-dimethyl-N'-(phenylmethyl)-N'-2-pyridinyl-, monohydrochloride;1,2-Ethanediamine, N1,N1-dimethyl-N2-(phenylmethyl)-N2-2-pyridinyl-, hydrochloride, Pyridine, 2-[benzyl[2-(dimethylamino)ethyl]amino]-, monohydrochloride, 2-[Benzyl[2-(dimethylamino)ethyl]amino]pyridine hydrochloride, Azaron, Dehistin, Dehistin monohydrochloride, N,N-Dimethyl-N'-(2-pyridyl)-N'-benzylethylenediamine hydrochloride, N-Benzyl-N',N'-dimethyl-N-2-pyridylethylenediamine hydrochloride, NIH 10186, PBZ, Piristin, Pyribenzamine hydrochloride, Pyribenzamine monohydrochloride, Pyrinamine, ReCovr, Stanzamine, Tripelennamine hydrochloride, Tripelennamine monohydrochloride, Vetibenzamine, Vetobenzamina
InChI formel InChI=1S/C16H21N3.ClH/c1-18(2)12-13-19(16-10-6-7-11-17-16)14-15-8-4-3-5-9-15;/h3-11H,12-14H2,1-2H3;1H
SMIL Cl.CN(C)CCN(Cc1ccccc1)c2ccccn2
IUPAC navn N'-benzyl-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;hydrochloride
Opløsningsmiddel eller Matrix Acetonitrile
Molekylvægt (g/mol) 291.82
Mængde 1 ml
Formel vægt 291.1502 g/mol