missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimipramine N-Oxide, TRC
Specifications
Specifications
| Kemisk navn eller materiale | (2RS)-3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine N-oxide (Trimipramine N-Oxide) |
| CAS | 14171-70-1 |
| Molekylær formel | C20 H26 N2 O |
| Mængde | 5 mg |
| Synonym | (2RS)-3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine N-oxide, Trimipramine Maleate Imp. A (EP), Trimipramine N-Oxide |
| InChI formel | InChI=1S/C20H26N2O/c1-16(15-22(2,3)23)14-21-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)21/h4-11,16H,12-15H2,1-3H3 |
| SMIL | CC(CN1c2ccccc2CCc3ccccc13)C[N+](C)(C)[O-] |
| IUPAC navn | 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N,2-trimethylpropan-1-amine oxide |
| Molekylvægt (g/mol) | 310.43 |
| Formel vægt | 310.2045 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?