Få mere at vide
Trimetozine, LoGiCal
Artikelnummer. 30462143
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462143 10 mg
1 options
Artikelnummer. 30462143 Leverandør LoGiCal Leverandørnr. LGCFOR3363.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trimetozine
CAS 635-41-6
Analyt- eller komponentnavne Trimetozine
Forsendelsestilstand Room Temperature
Molekylær formel C14 H19 N O5
Certificeringer/compliance ISO 9001
Synonym Methanone, 4-morpholinyl(3,4,5-trimethoxyphenyl)-, Morpholine, 4-(3,4,5-trimethoxybenzoyl)- (6CI,7CI,8CI,9CI), 4-(3,4,5-Trimethoxybenzoyl)morpholine, 4-(3,4,5-Trimethoxybenzoyl)tetrahydro-1,4-oxazine, Abbott 22370, N-(3,4,5-Trimethoxybenzoyl)tetrahydro-1,4-oxazine, NSC 62939, Opalene, PS 2383, Sedoxazine, Trifenoxazin, Trimethoxazine, Trimetozin, Trimetozine, Trioxazin, Trioxazine, V 7
InChI formel InChI=1S/C14H19NO5/c1-17-11-8-10(9-12(18-2)13(11)19-3)14(16)15-4-6-20-7-5-15/h8-9H,4-7H2,1-3H3
Løsningstype Neat
SMIL COc1cc(cc(OC)c1OC)C(=O)N2CCOCC2
IUPAC navn morpholin-4-yl-(3,4,5-trimethoxyphenyl)methanone
Industry Type Clinical Testing
Molekylvægt (g/mol) 281.3
Mængde 10 mg
Formel vægt 281.1263 g/mol